Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif1ax | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025437.4 | NP_079713.2 | 3 | 336 | ||
GenBank mRNA | AK005185.1 | - | 3 | 324 | ||
AK007301.1 | - | 3 | 287 | |||
AK012233.1 | - | 3 | 316 | |||
AK012464.1 | - | 3 | 237 | |||
AK012714.1 | - | 3 | 319 | |||
AK021283.1 | - | 3 | 317 | |||
AK030774.1 | - | 340 | ||||
AK031113.1 | - | 348 | ||||
AK090053.1 | - | 302 | ||||
AK134827.1 | - | 261 | ||||
AK134969.1 | - | 261 | ||||
AK165574.1 | - | 302 | ||||
AK166629.1 | - | 3 | 339 | |||
AK166852.1 | - | 3 | 337 | |||
AK187388.1 | - | 2 | 76 | |||
AK207164.1 | - | 2 | 61 | |||
AK210743.1 | - | 2 | 50 | |||
AK217305.1 | - | 2 | 98 | |||
BC027284.1 | - | 3 | 273 | 3156134 |