Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dimt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025447.4 | NP_079723.1 | 10 | 856 | ||
GenBank mRNA | AK007759.1 | - | 10 | 808 | ||
AK011544.1 | - | 10 | 813 | |||
AK015145.1 | - | 10 | 805 | |||
AK078186.1 | - | 8 | 1650 | |||
AK078401.1 | - | 8 | 1672 | |||
AK146188.1 | - | 10 | 827 | |||
AK161333.1 | - | 10 | 808 | |||
BC019799.1 | - | 10 | 829 | 5053500 |