Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klf15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023184.3 | NP_075673.1 | 2 | 405 | ||
GenBank mRNA | AF317225.1 | - | 1 | 280 | ||
AK009739.1 | - | 2 | 405 | |||
AK138824.1 | - | 2 | 442 | |||
AK142618.1 | - | 2 | 407 | |||
BC013486.1 | - | 1 | 325 | 4211067 |