Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cox14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183256.3 | NP_899079.1 | 2 | 459 | ||
GenBank mRNA | AK002816.1 | - | 2 | 322 | ||
AK004378.1 | - | 2 | 278 | |||
AK009401.1 | - | 2 | 431 | |||
BC059905.1 | - | 2 | 411 | 6823331 | ||
BC092142.1 | - | 2 | 360 | 6814201 | ||
BP774190.1 | - | 2 | 486 |