Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sln | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025540.2 | NP_079816.1 | 2 | 334 | ||
GenBank mRNA | AK008863.1 | - | 2 | 337 | ||
AK008896.1 | - | 2 | 335 | |||
AK009005.1 | - | 2 | 337 | |||
AK009809.1 | - | 422 | ||||
BC028496.1 | - | 2 | 315 | 1397989 | ||
CA460968.1 | - | 2 | 372 |