Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2E3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278554.1 | NP_001265483.1 | 4, 5 | 765 | ||
NM_001278555.1 | NP_001265484.1 | 4, 5 | 573 | |||
NM_006357.3 | NP_006348.1 | 4, 5 | 804 | |||
NM_182678.2 | NP_872619.1 | 5, 6 | 605 | |||
XM_005246244.2 | XP_005246301.1 | 4, 5 | 761 | |||
GenBank mRNA | AB017644.1 | - | 4, 5 | 447 | ||
AF085362.1 | - | 4, 5 | 505 | |||
AK314145.1 | - | 4, 5 | 775 | |||
BC003554.1 | - | 4, 5 | 487 | 3537280 | ||
BC092407.1 | - | 4, 5 | 744 | 6180242 | ||
BI257596.1 | - | 4, 5 | 567 | |||
BQ932163.1 | - | 5, 6 | 605 | |||
BT019345.1 | - | 3, 4 | 386 |
Gene Symbol: UBE2E4P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110506.1 | 1 | 389 | |||
GenBank mRNA | AF136176.1 | - | 1 | 389 |