Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pyurf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025574.3 | NP_079850.1 | 2 | 342 | ||
GenBank mRNA | AK003713.1 | - | 2 | 343 | ||
AK008978.1 | - | 2 | 342 | |||
AK011515.1 | - | 2 | 279 | |||
AK029193.1 | - | 318 | ||||
AK032381.1 | - | 2 | 335 | |||
BC029232.1 | - | 2 | 306 | 4164517 |