Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC9A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042537.1 | NP_001036002.1 | 7 | 1025 | ||
NM_001177651.1 | NP_001171122.1 | 8 | 903 | |||
NM_006359.2 | NP_006350.1 | 7 | 929 | |||
XM_006724726.3 | XP_006724789.1 | 8 | 1019 | |||
XM_011531243.2 | XP_011529545.1 | 8 | 783 | |||
XM_017029223.1 | XP_016884712.1 | 8 | 1036 | |||
XM_017029224.1 | XP_016884713.1 | 8 | 1166 | |||
XM_017029225.1 | XP_016884714.1 | 8 | 803 | |||
GenBank mRNA | AB074255.1 | - | 7 | 949 | ||
AF030409.1 | - | 7 | 888 | |||
AK289775.1 | - | 7 | 949 | |||
AK296444.1 | - | 7 | 688 | |||
AK297218.1 | - | 6 | 475 | |||
AK300475.1 | - | 8 | 894 | |||
AK314562.1 | - | 7 | 1017 | |||
AK316255.1 | - | 8 | 894 | |||
BC035029.1 | - | 7 | 987 | 5264865 | ||
BC049169.1 | - | 7 | 949 | 5312949 | ||
D87743.1 | - | 7 | 844 |