Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm10413 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029288.3 | NP_083564.2 | 1 | 128 | ||
GenBank mRNA | AK005583.1 | - | 1 | 118 | ||
BC100412.1 | - | 1 | 128 | 30916569 |
Gene Symbol: Gm5795 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270806.1 | NP_001257735.1 | 1 | 99 | ||
GenBank mRNA | BU946301.1 | - | 1 | 102 |