Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem41a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025693.4 | NP_079969.1 | 2 | 250 | ||
GenBank mRNA | AK007685.1 | - | 2 | 245 | ||
AK160292.1 | - | 2 | 170 | |||
BB853849.1 | - | 2 | 250 | |||
BB873279.1 | - | 2 | 250 | |||
BC011512.1 | - | 2 | 230 | |||
BC019770.1 | - | 2 | 205 | 5124769 |