Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006366.2 | NP_006357.1 | 10 | 1271 | ||
XM_011514233.2 | XP_011512535.1 | 9 | 1482 | |||
GenBank mRNA | AK293134.1 | - | 8 | 872 | ||
AK294215.1 | - | 4 | 333 | |||
AK295600.1 | - | 7 | 807 | |||
AK297538.1 | - | 9 | 987 | |||
AK308461.1 | - | 10 | 1090 | |||
AK312362.1 | - | 10 | 1399 | |||
AL832154.1 | - | 8 | 933 | |||
BC008481.1 | - | 10 | 1064 | 4295984 | ||
CR457022.1 | - | 9 | 1028 | |||
U02390.1 | - | 10 | 1111 |