Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105530.1 | NP_001099000.1 | 7 | 793 | ||
NM_006367.3 | NP_006358.1 | 7 | 811 | |||
XM_005270368.1 | XP_005270425.1 | 7 | 651 | |||
XM_011540509.1 | XP_011538811.1 | 8 | 1204 | |||
XM_011540510.1 | XP_011538812.1 | 8 | 1201 | |||
XM_011540511.1 | XP_011538813.1 | 7 | 635 | |||
XM_011540512.1 | XP_011538814.1 | 7 | 754 | |||
XM_011540513.2 | XP_011538815.1 | 7 | 646 | |||
XM_011540514.1 | XP_011538816.1 | 8 | 761 | |||
XM_011540515.1 | XP_011538817.1 | 8 | 1201 | |||
XM_017000067.1 | XP_016855556.1 | 7 | 719 | |||
XM_017000068.1 | XP_016855557.1 | 8 | 763 | |||
XM_017000069.1 | XP_016855558.1 | 8 | 974 | |||
XM_017000070.1 | XP_016855559.1 | 7 | 993 | |||
XM_017000071.1 | XP_016855560.1 | 7 | 951 | |||
XM_017000072.1 | XP_016855561.1 | 7 | 887 | |||
XM_017000073.1 | XP_016855562.1 | 7 | 708 | |||
GenBank mRNA | AK222513.1 | - | 7 | 652 | ||
AK295396.1 | - | 8 | 573 | |||
AK297829.1 | - | 7 | 520 | |||
AK298094.1 | - | 7 | 491 | |||
AK298110.1 | - | 7 | 595 | |||
AK300861.1 | - | 6 | 490 | |||
AK315731.1 | - | 7 | 698 | |||
BC013963.2 | - | 7 | 628 | 2963922 | ||
BC095440.1 | - | 7 | 633 | 30370256 | ||
BT007152.1 | - | 6 | 582 | |||
CR457409.1 | - | 6 | 582 | |||
L12168.1 | - | 6 | 621 | |||
M98474.1 | - | 7 | 652 |