Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006369.4 | NP_006360.3 | 4 | 457 | ||
XM_017000074.1 | XP_016855563.1 | 4 | 483 | |||
GenBank mRNA | AF370430.1 | - | 4 | 357 | ||
AK024051.1 | - | 4 | 366 | |||
AK125827.1 | - | 3 | 1734 | |||
AK291283.1 | - | 4 | 440 | |||
AK296436.1 | - | 4 | 276 | |||
AK297596.1 | - | 4 | 471 | |||
BC092479.1 | - | 4 | 267 | |||
BC101522.1 | - | 4 | 447 | 8069028 | ||
BC101524.1 | - | 4 | 447 | 8069030 | ||
BM548142.1 | - | 417 | ||||
BX537366.1 | - | 4 | 368 | |||
CF125098.1 | - | 4 | 458 |