Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mettl7a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081471.1 | NP_001074940.1 | 2 | 986 | ||
GenBank mRNA | AB253945.1 | - | 2 | 985 |
Gene Symbol: Mettl7a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027334.3 | NP_081610.2 | 2 | 1015 | ||
GenBank mRNA | AB253943.1 | - | 2 | 983 | ||
AJ536232.1 | - | 2 | 987 | |||
AK076169.1 | - | 2 | 1017 | |||
BC033446.1 | - | 2 | 992 | |||
BC047149.1 | - | 2 | 1000 | |||
BC055034.1 | - | 2 | 1006 | 6306873 | ||
BC060144.1 | - | 2 | 995 |
Gene Symbol: Mettl7a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199477.2 | NP_955771.2 | 2 | 995 | ||
GenBank mRNA | AK143923.1 | - | 2 | 996 | ||
BC087869.1 | - | 2 | 995 | 30304748 | ||
BC090971.1 | - | 2 | 995 | 30314995 |