Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006373.3 | NP_006364.2 | 3, 4 | 895 | ||
GenBank mRNA | AK097072.1 | - | 4, 5 | 722 | ||
AK290460.1 | - | 3, 4 | 871 | |||
AK298536.1 | - | 3, 4 | 605 | |||
AK315838.1 | - | 3, 4 | 459 | |||
BC001913.1 | - | 3, 4 | 764 | |||
BC008725.2 | - | 3, 4 | 819 | 3162543 | ||
BC014279.2 | - | 3, 4 | 801 | 3834206 | ||
BC015041.2 | - | 3, 4 | 833 | 3919901 | ||
BC090858.1 | - | 3, 4 | 833 | 6528247 | ||
BT007369.1 | - | 3, 4 | 775 | |||
U18009.1 | - | 3, 4 | 533 | |||
U25779.1 | - | 3, 4 | 500 |