Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem107 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025838.2 | NP_080114.1 | 5 | 558 | ||
NM_028336.3 | NP_082612.2 | 4 | 784 | |||
GenBank mRNA | AK002804.1 | - | 5 | 528 | ||
AK003403.1 | - | 5 | 554 | |||
AK012938.1 | - | 4 | 783 | |||
AK154279.1 | - | 5 | 556 | |||
BC096686.1 | - | 5 | 536 | 1446796 |