Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BNIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320674.1 | NP_001307603.1 | 3 | 544 | ||
NM_001320675.1 | NP_001307604.1 | 3 | 339 | |||
NM_004330.3 | NP_004321.2 | 3 | 544 | |||
GenBank mRNA | AK299628.1 | - | 3 | 339 | ||
AK308237.1 | - | 3 | 453 | |||
BC002461.2 | - | 3 | 329 | 3343559 | ||
U15173.1 | - | 3 | 298 |