Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENOX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281736.1 | NP_001268665.1 | 14 | 2034 | ||
NM_006375.3 | NP_006366.2 | 15 | 2178 | |||
NM_182314.2 | NP_872114.1 | 16 | 2267 | |||
XM_005262354.3 | XP_005262411.1 | 15 | 2123 | |||
XM_011531244.2 | XP_011529546.1 | 16 | 2269 | |||
XM_011531245.2 | XP_011529547.1 | 16 | 2263 | |||
XM_011531247.2 | XP_011529549.1 | 17 | 2395 | |||
XM_011531249.2 | XP_011529551.1 | 14 | 1823 | |||
XM_011531251.2 | XP_011529553.1 | 14 | 1928 | |||
XM_017029226.1 | XP_016884715.1 | 18 | 2486 | |||
XM_017029227.1 | XP_016884716.1 | 16 | 2220 | |||
XM_017029228.1 | XP_016884717.1 | 15 | 1969 | |||
XM_017029229.1 | XP_016884718.1 | 15 | 2172 | |||
GenBank mRNA | AF207881.2 | - | 13 | 1788 | ||
AK000353.1 | - | 14 | 1951 | |||
AK289812.1 | - | 16 | 2159 | |||
AK289837.1 | - | 15 | 2070 | |||
BC019254.1 | - | 10 | 1303 | |||
BC139781.1 | - | 13 | 1851 | |||
BC140874.1 | - | 14 | 1948 | 9021742 | ||
S72904.1 | - | 8 | 944 |