Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENOX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281736.1 | NP_001268665.1 | 12 | 1859 | ||
NM_006375.3 | NP_006366.2 | 13 | 2003 | |||
NM_182314.2 | NP_872114.1 | 14 | 2092 | |||
XM_005262354.3 | XP_005262411.1 | 13 | 1948 | |||
XM_011531244.2 | XP_011529546.1 | 14 | 2094 | |||
XM_011531245.2 | XP_011529547.1 | 14 | 2088 | |||
XM_011531247.2 | XP_011529549.1 | 15 | 2220 | |||
XM_011531249.2 | XP_011529551.1 | 12 | 1648 | |||
XM_011531251.2 | XP_011529553.1 | 12 | 1753 | |||
XM_017029226.1 | XP_016884715.1 | 16 | 2311 | |||
XM_017029227.1 | XP_016884716.1 | 14 | 2045 | |||
XM_017029228.1 | XP_016884717.1 | 13 | 1794 | |||
XM_017029229.1 | XP_016884718.1 | 13 | 1997 | |||
GenBank mRNA | AF207881.2 | - | 11 | 1613 | ||
AK000353.1 | - | 12 | 1776 | |||
AK289812.1 | - | 14 | 1984 | |||
AK289837.1 | - | 13 | 1895 | |||
BC019254.1 | - | 8 | 1128 | |||
BC139781.1 | - | 11 | 1676 | |||
BC140874.1 | - | 12 | 1773 | 9021742 | ||
S72904.1 | - | 6 | 769 |