Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENOX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281736.1 | NP_001268665.1 | 11 | 1761 | ||
NM_006375.3 | NP_006366.2 | 12 | 1905 | |||
NM_182314.2 | NP_872114.1 | 13 | 1994 | |||
XM_005262354.3 | XP_005262411.1 | 12 | 1850 | |||
XM_011531244.2 | XP_011529546.1 | 13 | 1996 | |||
XM_011531245.2 | XP_011529547.1 | 13 | 1990 | |||
XM_011531247.2 | XP_011529549.1 | 14 | 2122 | |||
XM_011531249.2 | XP_011529551.1 | 11 | 1550 | |||
XM_011531251.2 | XP_011529553.1 | 11 | 1655 | |||
XM_017029226.1 | XP_016884715.1 | 15 | 2213 | |||
XM_017029227.1 | XP_016884716.1 | 13 | 1947 | |||
XM_017029228.1 | XP_016884717.1 | 12 | 1696 | |||
XM_017029229.1 | XP_016884718.1 | 12 | 1899 | |||
GenBank mRNA | AF207881.2 | - | 10 | 1515 | ||
AK000353.1 | - | 11 | 1678 | |||
AK289812.1 | - | 13 | 1886 | |||
AK289837.1 | - | 12 | 1797 | |||
BC019254.1 | - | 7 | 1030 | |||
BC139781.1 | - | 10 | 1578 | |||
BC140874.1 | - | 11 | 1675 | 9021742 | ||
S72904.1 | - | 5 | 671 |