Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufa6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025987.3 | NP_080263.1 | 3 | 355 | ||
GenBank mRNA | AK002749.1 | - | 3 | 302 | ||
AK010988.1 | - | 3 | 304 | |||
AK012328.1 | - | 3 | 304 | |||
AV087995.1 | - | 2 | 92 | |||
BC048451.1 | - | 3 | 355 | 6773780 |