Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UNC13B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006377.3 | NP_006368.3 | 4 | 536 | ||
XM_011517684.2 | XP_011515986.1 | 4 | 554 | |||
XM_011517685.2 | XP_011515987.1 | 4 | 554 | |||
GenBank mRNA | AB209998.1 | - | 4 | 519 | ||
AF020202.1 | - | 4 | 468 | |||
AK296836.1 | - | 4 | 505 | |||
BC111781.1 | - | 4 | 424 | 40037278 | ||
BC111800.1 | - | 4 | 424 | 40037280 |