Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CARM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199141.1 | NP_954592.1 | 10, 11 | 1314 | ||
XM_005259708.4 | XP_005259765.1 | 10, 11 | 1280 | |||
XM_011527638.2 | XP_011525940.1 | 9, 10 | 1070 | |||
GenBank mRNA | AF055027.1 | - | 5, 6 | 489 | ||
AL833242.1 | - | 10, 11 | 1107 | |||
BC046240.1 | - | 611 |