Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCOA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321703.1 | NP_001308632.1 | 16 | 3590 | ||
NM_001321707.1 | NP_001308636.1 | 16 | 3436 | |||
NM_001321711.1 | NP_001308640.1 | 15 | 3311 | |||
NM_001321712.1 | NP_001308641.1 | 15 | 3228 | |||
NM_001321713.1 | NP_001308642.1 | 15 | 3331 | |||
NM_006540.3 | NP_006531.1 | 16 | 3518 | |||
XM_017012961.1 | XP_016868450.1 | 16 | 3505 | |||
XM_017012962.1 | XP_016868451.1 | 16 | 3529 | |||
XM_017012963.1 | XP_016868452.1 | 16 | 4742 | |||
XM_017012964.1 | XP_016868453.1 | 16 | 3797 | |||
XM_017012965.1 | XP_016868454.1 | 16 | 3462 | |||
XM_017012966.1 | XP_016868455.1 | 17 | 3726 | |||
XM_017012967.1 | XP_016868456.1 | 15 | 3440 | |||
XM_017012968.1 | XP_016868457.1 | 17 | 3527 | |||
XM_017012969.1 | XP_016868458.1 | 15 | 3558 | |||
XM_017012970.1 | XP_016868459.1 | 15 | 9977 | |||
XM_017012971.1 | XP_016868460.1 | 15 | 3689 | |||
XM_017012972.1 | XP_016868461.1 | 12 | 3020 | |||
GenBank mRNA | AK298528.1 | - | 4 | 603 | ||
BC114383.1 | - | 15 | 3385 | 40082387 | ||
X97674.1 | - | 16 | 3463 |