Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srpr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026130.1 | NP_080406.1 | 10 | 1338 | ||
GenBank mRNA | AK004967.2 | - | 10 | 1338 | ||
AK038017.1 | - | 10 | 1326 | |||
AK183434.1 | - | 1 | 19 | |||
BC012512.1 | - | 421 | ||||
BC021839.1 | - | 10 | 1321 | 4207633 |