Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sdhb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023374.3 | NP_075863.2 | 4 | 532 | ||
GenBank mRNA | AF095936.1 | - | 1 | 83 | ||
AK003052.1 | - | 4 | 426 | |||
AK003533.1 | - | 4 | 416 | |||
AK009660.1 | - | 4 | 426 | |||
AK153045.1 | - | 4 | 409 | |||
AK169252.1 | - | 4 | 423 | |||
AK169839.1 | - | 4 | 425 | |||
AK196072.1 | - | 2 | 189 | |||
BC013509.1 | - | 4 | 427 | 4225025 | ||
BC018580.1 | - | 4 | 391 | |||
BC051934.1 | - | 4 | 532 | 5688898 | ||
DQ403000.1 | - | 3 | 323 |