Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nmral1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290761.1 | NP_001277690.1 | 2 | 513 | ||
NM_001290762.1 | NP_001277691.1 | 2 | 513 | |||
NM_001290763.1 | NP_001277692.1 | 2 | 513 | |||
NM_026393.2 | NP_080669.1 | 2 | 513 | |||
GenBank mRNA | AK012340.1 | - | 2 | 269 | ||
AK078389.1 | - | 2 | 269 | |||
AK146521.1 | - | 268 | ||||
BC030039.1 | - | 2 | 216 | 1313824 |