Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr83 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026399.2 | NP_080675.2 | 8 | 894 | ||
GenBank mRNA | AK005790.1 | - | 8 | 834 | ||
AK020654.1 | - | 8 | 855 | |||
AY365434.1 | - | 8 | 894 | |||
BC019369.1 | - | 8 | 817 | 4014928 | ||
BC020047.1 | - | 1901 |