Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rprm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023396.5 | NP_075885.1 | 1 | 1023 | ||
GenBank mRNA | AB043586.1 | - | 1 | 1012 | ||
AK010465.1 | - | 1 | 1024 | |||
AK014769.1 | - | 1 | 988 | |||
BC030065.1 | - | 1 | 846 | 1434823 |