Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2810417H13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026515.2 | NP_080791.2 | 1, 2 | 155 | ||
GenBank mRNA | AK011090.1 | - | 1, 2 | 161 | ||
AK011645.1 | - | 1, 2 | 118 | |||
AK011835.1 | - | 1, 2 | 115 | |||
AK013105.1 | - | 1, 2 | 115 | |||
AK017673.1 | - | 1, 2 | 118 | |||
AK134432.1 | - | 1, 2 | 118 | |||
AY496943.1 | - | 1, 2 | 49 | |||
BC021413.1 | - | 1, 2 | 101 | 5006272 | ||
BC096393.1 | - | 1, 2 | 91 | 1397865 |