Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam133b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042501.1 | NP_001035966.1 | 5, 6 | 403 | ||
GenBank mRNA | AK008115.1 | - | 5, 6 | 404 | ||
BC147310.1 | - | 5, 6 | 398 | 9056359 | ||
BC147311.1 | - | 5, 6 | 398 | 9056381 | ||
CK791263.1 | - | 4, 5 | 221 |