Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdhb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024221.3 | NP_077183.1 | 8 | 841 | ||
GenBank mRNA | AK011810.1 | - | 805 | |||
AK014180.1 | - | 8 | 796 | |||
AK014918.1 | - | 8 | 840 | |||
AK153058.1 | - | 8 | 795 | |||
AK166631.1 | - | 8 | 843 | |||
AK167798.1 | - | 8 | 797 | |||
AK218654.1 | - | 3 | 380 | |||
BC002188.1 | - | 6 | 665 | |||
BC019512.1 | - | 8 | 814 | 4210204 | ||
BC094468.1 | - | 8 | 787 | 5011266 |