Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxl15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133694.2 | NP_598455.2 | 4 | 1257 | ||
GenBank mRNA | AK003032.1 | - | 4 | 980 | ||
AK145009.1 | - | 4 | 1217 | |||
AK172339.1 | - | 5 | 1154 | |||
BC016499.1 | - | 4 | 1232 | 4221584 |