Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp1r14a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026731.3 | NP_081007.2 | 1 | 128 | ||
GenBank mRNA | AF352573.1 | - | 1 | 136 | ||
AK003249.1 | - | 1 | 127 | |||
AY050672.1 | - | 1 | 127 | |||
BC010832.1 | - | 1 | 128 | 4216018 |