Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172400.3 | NP_765988.2 | 4 | 305 | ||
GenBank mRNA | AK003892.2 | - | 4 | 294 | ||
AK049526.1 | - | 4 | 298 | |||
AK088325.1 | - | 4 | 309 | |||
AK150933.1 | - | 4 | 300 | |||
AK168645.1 | - | 4 | 289 | |||
BC035539.1 | - | 4 | 264 | |||
BC057191.1 | - | 4 | 276 | |||
BC064732.1 | - | 4 | 284 | |||
BC066222.1 | - | 4 | 284 | 30459247 | ||
BC080760.1 | - | 4 | 284 | |||
BC137735.1 | - | 4 | 301 | 8860754 | ||
BC145655.1 | - | 4 | 301 | |||
BY754536.1 | - | 4 | 309 |