Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mospd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001254762.1 | NP_001241691.1 | 3 | 552 | ||
NM_030037.2 | NP_084313.1 | 4 | 705 | |||
GenBank mRNA | AK088731.1 | - | 3 | 543 | ||
BC003880.1 | - | 3 | 562 | 3496491 | ||
BC034167.1 | - | 4 | 690 | |||
BC039620.1 | - | 4 | 686 | 5051528 | ||
BE861793.1 | - | 3 | 179 | |||
BQ445267.1 | - | 2 | 118 | |||
CK617131.1 | - | 3 | 503 |