Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glrx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038592.1 | NP_001033681.1 | 4 | 521 | ||
NM_001038593.1 | NP_001033682.1 | 4 | 424 | |||
NM_001038594.1 | NP_001033683.1 | 4 | 363 | |||
NM_023505.2 | NP_075994.2 | 4 | 400 | |||
GenBank mRNA | AF276918.1 | - | 4 | 366 | ||
AF380337.1 | - | 4 | 464 | |||
AK005853.1 | - | 4 | 424 | |||
AK142654.1 | - | 4 | 364 | |||
BC065387.1 | - | 4 | 398 | |||
BC132031.1 | - | 4 | 443 | 40130308 | ||
BC132033.1 | - | 4 | 443 | 40130310 | ||
BC145303.1 | - | 4 | 486 | |||
BC145304.1 | - | 4 | 443 | 9053858 |