Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHERP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006387.5 | NP_006378.3 | 5 | 608 | ||
GenBank mRNA | AF536542.1 | - | 5 | 575 | ||
BC001552.2 | - | 5 | 566 | |||
BC021294.2 | - | 5 | 593 | 4053575 | ||
CN336425.1 | - | 5 | 607 | |||
U94836.2 | - | 4 | 510 |