Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nup43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145706.2 | NP_663752.2 | 7 | 894 | ||
GenBank mRNA | AK011422.1 | - | 7 | 859 | ||
AK012191.1 | - | 7 | 888 | |||
AK090019.1 | - | 7 | 857 | |||
AK146262.1 | - | 7 | 896 | |||
BC011060.1 | - | 7 | 882 | 3156436 | ||
BF607996.1 | - | 6 | 662 | |||
DV046453.1 | - | 367 |