Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mettl7a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027334.3 | NP_081610.2 | 1 | 184 | ||
GenBank mRNA | AB253943.1 | - | 1 | 155 | ||
AJ536232.1 | - | 1 | 158 | |||
AK008925.1 | - | 1 | 213 | |||
AK076169.1 | - | 1 | 186 | |||
BC033446.1 | - | 1 | 161 | |||
BC047149.1 | - | 1 | 168 | |||
BC055034.1 | - | 1 | 175 | 6306873 | ||
BC060144.1 | - | 1 | 163 | |||
BC171988.1 | - | 1 | 178 | 9054274 | ||
BC171989.1 | - | 1 | 178 | 9054275 |