Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mettl7a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027334.3 | NP_081610.2 | 2 | 1594 | ||
GenBank mRNA | AB253943.1 | - | 2 | 1562 | ||
AJ536232.1 | - | 2 | 1566 | |||
AK076169.1 | - | 2 | 1596 | |||
BC033446.1 | - | 2 | 1571 | |||
BC047149.1 | - | 2 | 1579 | |||
BC055034.1 | - | 2 | 1585 | 6306873 | ||
BC060144.1 | - | 2 | 1574 |