Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abhd17c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133722.2 | NP_598483.2 | 3 | 969 | ||
GenBank mRNA | AK211213.1 | - | 2 | 217 | ||
BC018511.1 | - | 3 | 969 | 3483379 | ||
BM943880.1 | - | 3 | 450 |