Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf126 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144528.3 | NP_653111.1 | 9 | 1336 | ||
GenBank mRNA | AK011369.1 | - | 1 | 213 | ||
AK042319.1 | - | 8 | 1133 | |||
AK154781.1 | - | 9 | 1302 | |||
BB823711.1 | - | 1 | 126 | |||
BC016543.1 | - | 9 | 1308 | 3158083 | ||
CF582846.1 | - | 1 | 344 |