Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fuz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027376.3 | NP_081652.2 | 9 | 1097 | ||
GenBank mRNA | AK033247.1 | - | 9 | 1084 | ||
AK035030.1 | - | 912 | ||||
AK045745.1 | - | 1092 | ||||
AK134650.1 | - | 9 | 1097 | |||
AK168939.1 | - | 8 | 1161 | |||
BC045601.1 | - | 7 | 2043 | |||
BC057127.1 | - | 2 | 825 | |||
CK624547.1 | - | 4 | 359 |