Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Basp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027395.2 | NP_081671.1 | 2 | 265 | ||
GenBank mRNA | AK011545.1 | - | 2 | 233 | ||
AK046868.1 | - | 2 | 239 | |||
BC063750.1 | - | 2 | 121 | |||
BC134388.1 | - | 1 | 47 |