Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Utp20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175158.3 | NP_780367.2 | 54 | 7243 | ||
GenBank mRNA | AB052760.1 | - | 12 | 1445 | ||
AK047081.1 | - | 1294 | ||||
BC005522.1 | - | 12 | 1465 | |||
BC048955.1 | - | 24 | 3108 |