Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Senp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172068.1 | NP_001165539.1 | 4 | 488 | ||
NM_001172069.1 | NP_001165540.1 | 3 | 397 | |||
NM_001172070.1 | NP_001165541.1 | 3 | 378 | |||
NM_001172071.1 | NP_001165542.1 | 3 | 395 | |||
NM_027838.3 | NP_082114.1 | 2 | 285 | |||
GenBank mRNA | AK018606.1 | - | 3 | 376 | ||
AK029856.1 | - | 2 | 275 | |||
AK049120.1 | - | 4 | 478 | |||
BC037443.1 | - | 2 | 257 | 3599077 | ||
CO431667.1 | - | 3 | 362 | |||
DV645349.1 | - | 2 | 285 |