Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrwd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027891.4 | NP_082167.2 | 6 | 904 | ||
GenBank mRNA | AK004717.1 | - | 4 | 1866 | ||
AK042370.1 | - | 3 | 2154 | |||
AK051910.1 | - | 6 | 852 | |||
AK084948.1 | - | 6 | 904 | |||
AK153976.1 | - | 6 | 875 | |||
AK154352.1 | - | 6 | 846 | |||
AK167020.1 | - | 6 | 853 | |||
AK172708.1 | - | 6 | 872 | |||
AK217505.1 | - | 2 | 205 | |||
BC048397.1 | - | 6 | 883 | 6396815 |