Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wfdc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027961.1 | NP_082237.1 | 3 | 540 | ||
GenBank mRNA | AK007299.1 | - | 3 | 405 | ||
BC116847.1 | - | 3 | 540 | 40126166 | ||
BC116875.1 | - | 3 | 540 | 40126194 |