Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cars2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024248.1 | NP_077210.1 | 6 | 671 | ||
GenBank mRNA | AK009937.1 | - | 6 | 633 | ||
AK038968.1 | - | 6 | 684 | |||
AK041962.1 | - | 6 | 645 | |||
AK054486.1 | - | 6 | 668 | |||
CX204497.1 | - | 6 | 672 |